missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Phosphate Dibasic Trihydrate, Honeywell Fluka™
BIOPHARMA

Product Code. 15650730 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
30 kg
Unit Size:
1kg
30kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15650730 30 kg 30kg
15640730 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15650730 Supplier Honeywell-Fluka Supplier No. 0426030KG
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss special, meets analytical specification of DAB,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-75-4
Molecular Formula H7MgO7P
Molecular Weight (g/mol) 174.33
MDL Number MFCD00150390
InChI Key OKIWLDVQGKRUNR-UHFFFAOYSA-L
Synonym magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
PubChem CID 16204530
SMILES O.O.O.[Mg++].OP([O-])([O-])=O

Specifications

Quantity 30 kg
Linear Formula MgHPO4 · 3H2O
UN Number NONH for all modes of transport
IUPAC Name magnesium(2+) trihydrate hydrogen phosphate
Formula Weight 174.33g/mol
Percent Purity ≥98%
Grade puriss. special
Density 2.123g/mL at 25°C
Chemical Name or Material Magnesium Phosphate Dibasic Trihydrate, Meets analytical specification of DAB
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.