missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Perchlorate, TRC

Product Code. 30478163
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478163 5 g 5g
30472863 25 g 25g
30469427 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478163 Supplier Toronto Research Chemicals Supplier No. TRCM1104755G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10034-81-8
Molecular Formula 2(ClO4) . Mg
Molecular Weight (g/mol) 223.2
Synonym Perchloric Acid, Magnesium Salt, Anhydron, Anhydrone, Anhydrous Magnesium Perchlorate, Dehydrite, KhKM 300, Magnesium Diperchlorate, Magnesium Perchlorate (Mg(ClO4)2), Magnesium(II) Perchlorate, XKM 300
IUPAC Name magnesium;diperchlorate
SMILES [Mg+2].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/2ClHO4.Mg/c2*2-1(3,4)5;/h2*(H,2,3,4,5);/q;;+2/p-2
Formula Weight 221.8821
Chemical Name or Material Magnesium Perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.