missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Perchlorate, Pure, Dried, Desiccating Agent, Fisher Chemical™

Product Code. p-7073282 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
Unit Size:
250g
This item is not returnable. View return policy

Product Code. 10254630

Brand: Fisher Chemical M/0950/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10034-81-8
Molecular Formula Cl2MgO8
Molecular Weight (g/mol) 223.197
MDL Number 11102
InChI Key MPCRDALPQLDDFX-UHFFFAOYSA-L
Synonym anhydrone, magnesium perchlorate, dehydrite, perchloric acid, magnesium salt, anhydrous magnesium perchlorate, unii-7n77z541yf, hsdb 661, magnesium diperchlorate, magnesium perchlorate, anhydrous, perchlorate de magnesium french
PubChem CID 24840
IUPAC Name magnesium;diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Specifications

Melting Point 250°C
Color White
pH 5 to 9
Physical Form Solid
Quantity 250 g
Formula Weight 223.21g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Magnesium perchlorate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.