missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Perchlorate Hydrate, Puriss. p.a., ≥99.0% (calc. based on dry substance, KT), Honeywell™ Fluka™

Product Code. 15696070 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15696070

Brand: Honeywell-Fluka 63100500G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a.,≥99.0% (calc. based on dry substance, KT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 64010-42-0
Color White
Physical Form Powder
Quantity 500 g
Molecular Formula Cl2H2MgO9
Linear Formula Mg(ClO4)2 · xH2O
MDL Number MFCD00149776
UN Number UN1475
Synonym magnesium perchlorate hydrate, magnesium diperchlorate hydrate, 2clo4.mg.h2o, magnesium 2+ ion hydrate diperchlorate, magnesium 2+ hydrate diperchlorate, magnesium perchlorate hydrate, puriss. p.a calc. based on dry substance, kt
InChI Key KKFFNQXCJMNXHI-UHFFFAOYSA-L
SMILES O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]
IUPAC Name magnesium;diperchlorate;hydrate
Molecular Weight (g/mol) 241.212
PubChem CID 6915996
Formula Weight 223.21g/mol (anhydrous basis)
Percent Purity ≥99.0% (calc. based on dry substance, KT)
Packaging Poly bottle
Chemical Name or Material Magnesium perchlorate hydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.