missing translation for 'onlineSavingsMsg'
Learn More

Magnesium perchlorate hexahydrate, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048839
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
13446-19-0
Cl2H12MgO14
331.287
MFCD00149778
MGBDNCBRZIMSDZ-UHFFFAOYSA-L
magnesium perchlorate hexahydrate, acmc-20akl9, 2clo4.mg.6h2o, ksc491a3f, magnesium 2+ ion hexahydrate diperchlorate, magnesium 2+ hexahydrate diperchlorate
16211969
magnesium;diperchlorate;hexahydrate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]
This item is not returnable. View return policy
13446-19-0
Cl2H12MgO14
331.287
MFCD00149778
MGBDNCBRZIMSDZ-UHFFFAOYSA-L
magnesium perchlorate hexahydrate, acmc-20akl9, 2clo4.mg.6h2o, ksc491a3f, magnesium 2+ ion hexahydrate diperchlorate, magnesium 2+ hexahydrate diperchlorate
16211969
magnesium;diperchlorate;hexahydrate
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Mg+2]

Commercially available magnesium perchlorate is reported as an extremely efficient catalyst for the synthesis of α-aminophosphonates. Magnesium perchlorate has been found to be an efficient catalyst for the synthesis of imines and phenylhydrazones by the reaction of carbonyl compounds with amines and phenylhydrazine in high yields at room temperatures and in short times.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 185°C to 190°C
Color White
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99% (Metals basis)
UN Number UN1475
Merck Index 14,5678
Solubility Information Insoluble in water.
Formula Weight 331.33 (223.23 Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Magnesium perchlorate hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'