missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E 530, light, 98.0-100.5% (calc. for dried substance), Honeywell Fluka™
BIOPHARMA

Product Code. 15611250 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
20 kg
Unit Size:
1kg
20kg
500g
This item is not returnable. View return policy

Product Code. 15611250

Brand: Honeywell-Fluka 1313820KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E 530, light, 98.0-100.5% (calc. for dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Boiling Point 3,600°C
Melting Point 2852°C (literature)
CAS Max % 100.0000%
Quantity 20 kg
Assay Percent Range 98.0 to 100.5%
Linear Formula MgO
UN Number NONH for all modes of transport
Formula Weight 40.3
Percent Purity 98.0 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Paper / Poly Bag
Density 3.58 g/cm3
Chemical Name or Material Magnesium oxide, Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E 530, light, 98.0-100.5% (calc. for dried substance)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.