missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Oxide, Extra Pure, Heavy, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™

Catalog Number 10244630
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
1309-48-4
MgO
40.30
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
14792
oxomagnesium
O=[Mg]
This item is not returnable. View return policy
1309-48-4
MgO
40.30
MFCD00011109
CPLXHLVBOLITMK-UHFFFAOYSA-N
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
14792
oxomagnesium
O=[Mg]
Boiling Point 3600°C
Melting Point 2800°C
Color White
pH 10.3
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 40.29g/mol
Packaging Plastic powder jar
Vapor Pressure 0 hPa at 20°C
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'