missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 99.95% (metals basis)

Product Code. 11386988
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386988 25 g 25g
11396988 100 g 100g
11376988 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11386988 Supplier Thermo Scientific Alfa Aesar Supplier No. 014684.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Vacuum Deposition | CAS: 1309-48-4 | MgO | 40.30 g/mol

Magnesium oxide is used as a refractive material, which is physically and chemically stable at higher temperature. Magnesium oxide is a raw material for producing portland cement. It is also used in agricultural, chemical construction, environment al and other industrial applications. It is also used in medicine, colorimetry, industrial cables, heating elements, ceramics, spin-tunneling devices, and several other materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Boiling Point 3600°C
Melting Point 2852°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.95% (Metals basis)
Solubility Information Soluble in acid and ammonia. Insoluble in alcohol.
Formula Weight 40.3
Refractive Index 1.736
Density 3.58 g/cm3 at 20°C
Chemical Name or Material Magnesium oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.