missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 99.95% (metals basis)

Product Code. p-7076403
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11348788

Brand: Thermo Scientific Alfa Aesar 041648.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used as a laboratory reagent, and a reference white color for colorimetry. Magneisum oxide microspheres is used for enrichment method of polycyclic aromatic hydrocarbons. It is also used in groundwater studies. | CAS: 1309-48-4 | MgO | 40.30 g/mol

Magnesium oxide is used as a laboratory reagent, and a good reference white color for colorimetry. Magneisum oxide microspheres is used for enrichment method of polycyclic aromatic hydrocarbons. It is also used in groundwater studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Boiling Point 3600°C
Melting Point 2852°C
Physical Form Fused Lump
Quantity 25 g
Solubility Information Soluble in HCl and dilute acids. Insoluble in water and ethanol.
Formula Weight 40.3
Percent Purity 99.95%
Packaging Glass bottle
Refractive Index 1.736
Specific Gravity 3.58
Density 3.58
Chemical Name or Material Magnesium oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.