missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Oxide, 99.9% Trace metals basis, Honeywell™

Product Code. 15634150 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 15634150

Brand: Honeywell Research Chemicals 342777100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 3600°C (1013hPa)
Melting Point 2852°C
CAS 1309-48-4
Color White
pH Alkaline
Physical Form Powder
Quantity 100 g
Molecular Formula MgO
Linear Formula MgO
MDL Number MFCD00011109
UN Number NONH for all modes of transport
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
Solubility Information Practically insoluble in water
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES O=[Mg]
IUPAC Name oxomagnesium
Molecular Weight (g/mol) 40.30
PubChem CID 14792
Formula Weight 40.3g/mol
Percent Purity 99.9% (trace metals basis)
Odor Odorless
Packaging Poly bottle
Density ∽3.58g/cu.cm at 20°C
Chemical Name or Material Magnesium oxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.