missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 95+%, ACS reagent

Product Code. 10157143 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10157143 250 g Glass bottle 250g
10712602 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157143 Supplier Thermo Scientific Acros Supplier No. 423892500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1309-48-4 | MgO | 40.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Boiling Point 3600°C
Melting Point 2800°C
Color White
Physical Form Granular Powder
Quantity 250 g
Assay Percent Range 95% min. (dried basis) (Complexometry)
Linear Formula MgO
Fieser 01,633; 02,256; 07,221; 16,200
Merck Index 15, 5741
Solubility Information Solubility in water: 6.2mg/l (20°C),reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids,insoluble in alcohol
Formula Weight 40.29
Percent Purity ≥95%
Grade ACS Reagent
Loss on Ignition 2.0% max.
Packaging Glass bottle
Specific Gravity 3.58
Density 3.5800g/mL
Chemical Name or Material Magnesium oxide, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.