missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxalate dihydrate, 98.5+%

Product Code. 11395028
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395028 100 g 100g
11385028 500 g 500g
11375028 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11395028 Supplier Thermo Scientific Alfa Aesar Supplier No. 043976.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 547-66-0 | C2H2MgO4+2 | 114.339 g/mol

Magnesium oxalate dihydrate is in the synthesis of nano sized magnesium oxide. It is also used as a catalyst, refractory materials, adsorbents, superconductors, and ferroelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-66-0
Molecular Formula C2H2MgO4+2
Molecular Weight (g/mol) 114.339
MDL Number MFCD00287279
InChI Key UHNWOJJPXCYKCG-UHFFFAOYSA-N
Synonym magnesium oxalate, magnesium oxalate 1:1, magnesium oxalate mgc2o4, oxalic acid, magnesium salt 1:1, ethanedioic acid, magnesium salt 1:1
PubChem CID 54611841
IUPAC Name magnesium;oxalic acid
SMILES C(=O)(C(=O)O)O.[Mg+2]

Specifications

Melting Point 150°C (decomposition)
Physical Form Powder, Pieces
Quantity 100 g
UN Number UN3288
Merck Index 14,5676
Solubility Information Soluble in diluted mineral acid,water (0.038g/100g H2O ). Insoluble in organics solutions.
Formula Weight 148.36 (112.32 Anhydrous)
Percent Purity ≥98.5%
Odor Odorless
Chemical Name or Material Magnesium oxalate dihydrate
compliance-icons
Product Identifier
  • Magnesium oxalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Magnesium oxalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.