missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitrate hydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11386897
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 11386897

Brand: Thermo Scientific Alfa Aesar 010799.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10377-60-3 | MgN2O6 | 148.31 g/mol

It is used in printing, chemical, agriculture and ceramics industries. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-60-3
Molecular Formula MgN2O6
Molecular Weight (g/mol) 148.31
MDL Number MFCD00011103
InChI Key YIXJRHPUWRPCBB-UHFFFAOYSA-N
Synonym magnesium nitrate, magnesium dinitrate, nitric acid, magnesium salt, unii-77cbg3un78, magnesium nitrate solution, nitric acid, magnesium salt 2:1, magniosan, hsdb 660, magnesium ii nitrate 1:2, magnesium standard for ic
PubChem CID 25212
ChEBI CHEBI:64736
IUPAC Name magnesium;dinitrate
SMILES [Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 330°C (decomposition)
Melting Point 89°C
Physical Form Crystalline Aggregates
Quantity 500 g
UN Number UN1474
Merck Index 14,5674
Solubility Information Freely soluble in water. Moderately soluble in ethanol and ammonia.
Formula Weight 148.32 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Sensitivity Hygroscopic
Density 1.46 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium nitrate hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.