missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Nitrate Hexahydrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10667222 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10667222 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10667222 Supplier Fisher Chemical Supplier No. M/0750/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point 330°C
Melting Point 89°C
CAS 13446-18-9
Color White
pH 5.0 to 8.2
Physical Form Solid
Quantity 500 g
Molecular Formula H12MgN2O12
MDL Number MFCD00149779
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name magnesium;dinitrate;hexahydrate
Molecular Weight (g/mol) 256.40
PubChem CID 202877
Formula Weight 256.4g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.