missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Nitrate Hexahydrate, Certified AR for Analysis, 99.0-101.0%, Fisher Chemical™

Product Code. 10755721 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10755721 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10755721 Supplier Fisher Chemical Supplier No. M/0752/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
IUPAC Name magnesium;dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Boiling Point 330°C
Melting Point 89°C
Color White
pH 5.0 to 8.2
Physical Form Solid
Quantity 500 g
Formula Weight 256.4g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.