missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Nitrate Hexahydrate, ≥98%, Honeywell Fluka™, BioXtra

Product Code. 15697950 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15697950 1 kg Poly bottle 1kg
15607960 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15697950 Supplier Honeywell-Fluka Supplier No. M52961KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BioXtra,≥98%

  • Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
  • Includes application-specific formulations, such as tablets and prepared solutions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 89°C
Color Colorless
Physical Form Solid
Quantity 1 kg
Linear Formula Mg(NO3)2 · 6H2O
UN Number NONH for all modes of transport
IUPAC Name magnesium;dinitrate;hexahydrate
Formula Weight 256.41g/mol
Percent Purity ≥98%
Odor Odorless
Packaging Poly bottle
Density 1.464g/cm3; Vapor Density: 6 (vs air)
Chemical Name or Material Magnesium nitrate hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.