missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Nitrate Hexahydrate, ≥98%, Honeywell Fluka™, BioXtra

Product Code. 15607960 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15607960 500 g Poly bottle 500g
15697950 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15607960 Supplier Honeywell-Fluka Supplier No. M5296500G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

BioXtra,≥98%

  • Stringent testing protocols may include trace metal analysis and specialized suitability testing, such as hybridoma or mouse embryo
  • Includes application-specific formulations, such as tablets and prepared solutions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-18-9
Molecular Formula H12MgN2O12
Molecular Weight (g/mol) 256.40
MDL Number MFCD00149779
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
PubChem CID 202877
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 89°C
Color Colorless
Physical Form Solid
Quantity 500 g
Linear Formula Mg(NO3)2 · 6H2O
UN Number NONH for all modes of transport
IUPAC Name magnesium(2+) hexahydrate dinitrate
Formula Weight 256.41g/mol
Percent Purity ≥98%
Odor Odorless
Packaging Poly bottle
Density 1.464g/cm3; Vapor Density: 6 (vs air)
Chemical Name or Material Magnesium nitrate hexahydrate
Show More Show Less
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.