missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitrate hexahydrate, 98+%, ACS reagent, ACROS Organics™

Product Code. 10127990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10127990 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10127990 Supplier Thermo Scientific Acros Supplier No. 423880050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point 330.0°C
Melting Point 89.0°C
CAS 13446-18-9
Color White
pH 5.0 to 8.2 (5% soln. at 25°C)
Physical Form Adhering Crystalline Powder
Quantity 5 g
Assay Percent Range 98+%
Molecular Formula H12MgN2O12
Linear Formula Mg(NO3)2·6H2O
MDL Number MFCD00149779
Flash Point 93.3°C
Merck Index 15, 5738
Synonym magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
Solubility Information Solubility in water: 420g/L (20°C). Other solubilities: freely soluble in alcohol, liquid nh3
InChI Key MFUVDXOKPBAHMC-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
IUPAC Name magnesium(2+) hexahydrate dinitrate
Molecular Weight (g/mol) 256.40
PubChem CID 202877
Formula Weight 256.4
Percent Purity ≥98%
Grade ACS Reagent
Packaging Glass bottle
Density 1.63
Chemical Name or Material Magnesium nitrate hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.