missing translation for 'onlineSavingsMsg'
Learn More

Magnesium nitrate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog Number 11404103
Click to view available options
Quantity:
250 g
1000 g
5000 g
13446-18-9
H12MgN2O12
256.40
MFCD00149779
MFUVDXOKPBAHMC-UHFFFAOYSA-N
magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
202877
magnesium;dinitrate;hexahydrate
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
13446-18-9
H12MgN2O12
256.40
MFCD00149779
MFUVDXOKPBAHMC-UHFFFAOYSA-N
magnesium nitrate hexahydrate, magnesium dinitrate hexahydrate, unii-v85k20ljmk, dusicnan horecnaty czech, v85k20ljmk, nitric acid, magnesium salt, hexahydrate, magnesium ii nitrate 1:2 , hexahydrate, magnesium 2+ ion hexahydrate dinitrate, magnesium, reference standard solution, dusicnan horecnaty
202877
magnesium;dinitrate;hexahydrate
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

Solubility
Very soluble in water, alcohol and liquid ammonia.

Notes
Heating of magnesium nitrate hexahydrate does not lead to dehydration but decomposition to produce magnesium oxide, nitrogen oxide and oxygen.
TRUSTED_SUSTAINABILITY
Boiling Point 330°C (decomposition)
Melting Point 89°C
Quantity 250 g
UN Number UN1474
Merck Index 14,5674
Solubility Information Very soluble in water,alcohol and liquid ammonia.
Formula Weight 256.41 (148.32 Anhydrous)
Percent Purity 98%
Sensitivity Hygroscopic
Density 1.46
Chemical Name or Material Magnesium nitrate hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'