missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium nitrate hexahydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.
Solubility
Very soluble in water, alcohol and liquid ammonia.
Notes
Heating of magnesium nitrate hexahydrate does not lead to dehydration but decomposition to produce magnesium oxide, nitrogen oxide and oxygen.
It is used in printing, chemical, agriculture and the ceramic industry. It is also used in the detection of selenium in plasma samples and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.
Solubility
Very soluble in water, alcohol and liquid ammonia.
Notes
Heating of magnesium nitrate hexahydrate does not lead to dehydration but decomposition to produce magnesium oxide, nitrogen oxide and oxygen.
Specifications
Boiling Point | 330°C (decomposition) |
Melting Point | 89°C |
Quantity | 1000 g |
UN Number | UN1474 |
Merck Index | 14,5674 |
Solubility Information | Very soluble in water,alcohol and liquid ammonia. |
Formula Weight | 256.41 (148.32 Anhydrous) |
Percent Purity | 98% |
Sensitivity | Hygroscopic |
Density | 1.46 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'