missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Methoxide Solution 6-10 wt. % in Methanol, TRC

Product Code. 30464325
Change view
Click to view available options
Quantity:
200 g
Unit Size:
200g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464325 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464325 Supplier Toronto Research Chemicals Supplier No. TRCM109880200G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Magnesium Methoxide
CAS 109-88-6
Molecular Formula 2(CH4O). Mg
Quantity 200 g
Synonym Magnesium Methylate, Bis(methoxy)magnesium, Dimethoxymagnesium, Magnesium Dimethoxide, Methanol, Magnesium Salt, Magnesium Methoxide solution 6-10 wt. % in Methanol
InChI Formula InChI=1S/2CH3O.Mg/c2*1-2;/h2*1H3;/q2*-1;+2
SMILES C[O-].[O-]C.[Mg+2]
IUPAC Name magnesium methanolate
Molecular Weight (g/mol) 88.38
Formula Weight 86.02
Concentration or Composition (by Analyte or Components) 45936 %
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.