missing translation for 'onlineSavingsMsg'
Learn More

Magnesium methoxide, 7-8% in methanol

Product Code. 11378408
Change view
Click to view available options
Quantity:
250 g
1 kg
2 kg
Unit Size:
1kg
250g
2kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378408 1 kg 1kg
11368408 250 g 250g
15411667 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378408 Supplier Thermo Scientific Alfa Aesar Supplier No. 040448.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MgO films | CAS: 109-88-6

The use of magnesium methoxide for the deprotection of alkyl esters is described. This mild reagent provides a good method to cleave esters efficiently and more importantly, allows for effective differentiation between two different esters. Dimethyl carbonate is synthesized directly from carbon dioxide and methanol in the presence of magnesium methoxide. 20 mol% DEA additions to magnesium methoxide showed enhanced stability such that a time-dependent change in the sol was not observed in air. Also enhanced the crystallization process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-88-6
MDL Number MFCD00053671

Specifications

Color Colorless to Yellow
Density 0.815 g/mL
Flash Point 14°C (57°F)
Odor Alcohol-like
Refractive Index 1.338
Quantity 1 kg
UN Number UN1230
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 86.38
Concentration or Composition (by Analyte or Components) 7 to 8% in methanol
Physical Form Liquid
Chemical Name or Material Magnesium methoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.