missing translation for 'onlineSavingsMsg'
Learn More

Magnesium meso-tetraphenylporphine monohydrate

Product Code. 11343728
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343728 2 g 2g
11353728 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343728 Supplier Thermo Scientific Alfa Aesar Supplier No. 039316.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14640-21-2 | C44H28MgN4 | 637.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14640-21-2
Molecular Formula C44H28MgN4
Molecular Weight (g/mol) 637.04
MDL Number MFCD00049815
InChI Key XEHJAWQTIIXDON-QHNUXLDVSA-N
Synonym magnesium meso-tetraphenylporphine hydrate
PubChem CID 131675734
IUPAC Name magnesium;2,7,12,17-tetraphenylporphyrin-21,22-diide
SMILES C1=CC=C(C=C1)C2=CC3=CC4=C(C=C([N-]4)C=C5C(=CC(=N5)C=C6C(=CC(=N6)C=C2[N-]3)C7=CC=CC=C7)C8=CC=CC=C8)C9=CC=CC=C9.[Mg+2]

Specifications

Quantity 2 g
Formula Weight 655.05 (637.04 Anhydrous)
Physical Form Crystalline
Chemical Name or Material Magnesium meso-tetraphenylporphine monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.