missing translation for 'onlineSavingsMsg'
Learn More

Magnesium lactate trihydrate, 99%

Product Code. 11378128
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378128 500 g 500g
11368128 25 g 25g
11388128 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378128 Supplier Thermo Scientific Alfa Aesar Supplier No. 039369.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 18917-96-9 | C6H16MgO9 | 256.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18917-96-9
Molecular Formula C6H16MgO9
Molecular Weight (g/mol) 256.49
MDL Number MFCD00156534
Synonym magnesium lactate trihydrate, c6h10mgo6.3h2o, 2-hydroxypropanoyl oxy magnesio 2-hydroxypropanoate trihydrate
PubChem CID 91668363
IUPAC Name magnesium;2-hydroxypropanoate;trihydrate

Specifications

Melting Point 41°C (decomposition)
Color White
Quantity 500 g
Merck Index 14,5672
Solubility Information Soluble in water, Slightly soluble in alcohol
InChI Key SNLQXUYQWUDJLB-UHFFFAOYNA-L
SMILES O.O.O.[Mg++].CC(O)C([O-])=O.CC(O)C([O-])=O
Formula Weight 256.49 (202.45 Anhydrous)
Percent Purity 99%
Physical Form Powder or Lumps
Chemical Name or Material Magnesium lactate trihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.