missing translation for 'onlineSavingsMsg'
Learn More

Magnesium iodide, ultra dry, 99.996% (metals basis)

Product Code. p-4781984
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11317867

Brand: Thermo Scientific Alfa Aesar 014283.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Light Sensitive and Hygroscopic | CAS: 10377-58-9 | I2Mg | 278.11 g/mol

Magnesium iodide is used as reagents and intermediates in pharmaceuticals. It is useful in Baylis-Hillman reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10377-58-9
Molecular Formula I2Mg
Molecular Weight (g/mol) 278.11
MDL Number MFCD00049485
InChI Key BLQJIBCZHWBKSL-UHFFFAOYSA-L
IUPAC Name magnesium(2+) diiodide
SMILES [Mg++].[I-].[I-]

Specifications

Melting Point 637°C (decomposition)
Color White
Physical Form Beads
Quantity 1 g
Assay Percent Range 99.996% (Metals basis)
Solubility Information Soluble in water(1480g/L at 18 C)
Formula Weight 278.12
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 4.43g/cm3 at 20°C
Chemical Name or Material Magnesium iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.