missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Hydroxide, TRC

Product Code. 30008847
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008847 25 g 25g
30006597 100 g 100g
30007194 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008847 Supplier Toronto Research Chemicals Supplier No. TRCM11036025G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2 Mg O2
Molecular Weight (g/mol) 58.32
Synonym Magnesium hydroxide, Magnesium Hydroxide (8CI), 10A, 10A (Hydroxide), 1110F, 200-06H, 2M-C01, 2N, 2N (Magnesium Hydroxide), 3320HT, 5D12, 936A, 96HS, Alcanex NHC 25, Apymag 40S, Apymag 80S, Apymag AOH 820, Aquamag, Aquamag (hydroxide), Asahi Glass 200-06, B 54, Burusaito, Combustrol 500, DG 800, DP 393, DSB 100, Daimushew 6000, Delamin FRA-B 15, Duhor, EP 1, FD 905U, FK 621, Finemag MO, Kisuma, Kyowamag F, M 3C2, Maglux ST, Magnesium Dihydroxide, Magnesium(II) Hydroxide, Magnifin A, Magseeds EP 2A, NF 2000, Nittomagu S, Novadol, Phillips Magnesia Tablets, Phillips Milk of Magnesia Liquid, Polysafe MG 2, Pural MG 100, QM 70, S 40 (hydroxide), S/G 84, SP, SP (hydroxide), SX 30MS;
IUPAC Name dihydroxymagnesium
SMILES O[Mg]O

Specifications

Quantity 25 g
InChI Formula InChI=1S/Mg.2H2O/h;2*1H2/q+2;;/p-2
Formula Weight 57.9905
Chemical Name or Material Magnesium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.