missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Hydroxide, 95%, Reagent grade, Honeywell™

Product Code. 15653160 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 kg
Packaging:
Poly Bottle
Unit Size:
25g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15653160 500 g Poly Bottle 500g
15633160 25 g Poly Bottle 25g
15643160 2 kg Poly Bottle 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15653160 Supplier Honeywell Research Chemicals Supplier No. 310093500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 95%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
PubChem CID 73981
ChEBI CHEBI:6637
IUPAC Name magnesium;dihydroxide
SMILES [OH-].[OH-].[Mg++]

Specifications

Melting Point 350°C
Quantity 500 g
Linear Formula Mg(OH)2
UN Number NONH for all modes of transport
Formula Weight 58.32g/mol
Percent Purity 95%
Grade Reagent
Packaging Poly Bottle
Density 2.39g/cm3
Chemical Name or Material Magnesium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.