missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hydroxide, 95%, pure

Product Code. p-44456 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10521574

Brand: Thermo Scientific Acros 315720250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1309-42-8 | H2MgO2 | 58.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
PubChem CID 73981
ChEBI CHEBI:6637
SMILES [OH-].[OH-].[Mg++]

Specifications

Melting Point 350°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 94% min. (Complexometry)
Linear Formula Mg(OH)2
Merck Index 15, 5734
Solubility Information Solubility in water: insoluble
IUPAC Name magnesium;dihydroxide
Formula Weight 58.33
Percent Purity 95%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Magnesium hydroxide, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.