missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hydroxide, 95-100.5%

Product Code. 11305929
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305929 100 g 100g
11331759 500 g 500g
11315929 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11305929 Supplier Thermo Scientific Alfa Aesar Supplier No. 012367.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Flame Retardant in Plastics, Such As PVC, As It Produces Less Toxic Fumes | CAS: 1309-42-8 | H2MgO2 | 58.32 g/mol

It is employed as an additive in certain refractories. It is applied as a non-hazardous alkali in neutralizing acidic wastewater. It is also used in making artificial reefs. It widely used in milk of magnesia antacids and laxatives. Also works as an antiperspirant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-42-8
Molecular Formula H2MgO2
Molecular Weight (g/mol) 58.32
MDL Number MFCD00011104
InChI Key VTHJTEIRLNZDEV-UHFFFAOYSA-L
Synonym milk of magnesia, magnesium dihydroxide, brucite, magnesium hydroxide suspension, magnesium hydroxide gilumag, magnesiumhydroxid, magnesiumhydroxide, magnesia,, acmc-1bqga, magnesium 2+ hydroxide
PubChem CID 73981
ChEBI CHEBI:6637
SMILES [OH-].[OH-].[Mg++]

Specifications

Melting Point 360°dec
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 95-100.5%
Solubility Information Soluble in dilute acids. Slightly soluble in water.
IUPAC Name magnesium;dihydroxide
Formula Weight 58.33
Odor Odorless
Sensitivity Air Sensitive
Appearance White powder
Density 2.36
Chemical Name or Material Magnesium hydroxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.