missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals

Product Code. 10339100 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10339100 5 g Glass bottle 5g
10608391 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10339100 Supplier Thermo Scientific Acros Supplier No. 264780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-75-4
Molecular Formula H7MgO7P
Molecular Weight (g/mol) 174.33
MDL Number MFCD00150390
InChI Key OKIWLDVQGKRUNR-UHFFFAOYSA-L
Synonym magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
PubChem CID 16204530
IUPAC Name magnesium;hydrogen phosphate;trihydrate
SMILES O.O.O.[Mg++].OP([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula MgHPO4·3H2O
Merck Index 14, 5681
Solubility Information Solubility in water: 0.25g/L (20°C). Other solubilities: soluble in diluted acids
Formula Weight 174.34
Percent Purity 98%
Grade Extra Pure
Packaging Glass bottle
Specific Gravity 2.13
Density 2.1300g/mL
Chemical Name or Material Magnesium hydrogen phosphate trihydrate, c.p., 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.