missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hexafluorosilicate hexahydrate 96%, ACROS Organics™

Product Code. 10464645 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
25g; Plastic bottle
This item is not returnable. View return policy

Product Code. 10464645

Brand: Thermo Scientific Acros 384670250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 18972-56-0
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 96%
Molecular Formula F6MgSi·6H2O
MDL Number MFCD00150250
Merck Index 15, 5732
Formula Weight 274.47
Packaging Plastic bottle
Chemical Name or Material Magnesium hexafluorosilicate hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium hexafluorosilicate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.