missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium foil, 0.25mm (0.01in) thick, hard, 99.9% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 100 x 100 mm |
| Assay Percent Range | 99.9% (metals basis) |
| Chemical Name or Material | Magnesium foil |
Safety and Handling
Product Identifier
- Magnesium foil
- Danger
- Flammable solid Category 1
- Self-heating substance Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H228-Flammable solid.
- H251-Self-heating: may catch fire.
- H261-In contact with water releases flammable gases.
- P223-Do not allow contact with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P420-Store away from other materials.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?