missing translation for 'onlineSavingsMsg'
Learn More

Magnesium fluoride, Optical Grade, 99.9% (metals basis)

Product Code. 11391628
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
100µL
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391628 10 g 10g
11533431 10 g 100µL
11301638 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11391628 Supplier Thermo Scientific Alfa Aesar Supplier No. 011021.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-40-6 | F2Mg | 62.30 g/mol

Magnesium fluoride finds application in optics, that are also used in space telescopes. It is also used in the electrolysis of aluminum ore and anti-reflective coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-40-6
Molecular Formula F2Mg
Molecular Weight (g/mol) 62.30
MDL Number MFCD00011108
InChI Key ORUIBWPALBXDOA-UHFFFAOYSA-L
IUPAC Name magnesium(2+) difluoride
SMILES [F-].[F-].[Mg++]

Specifications

Melting Point 1261°C
Color White
Physical Form Broken Crystals
Quantity 10 g
Merck Index 14,5665
Solubility Information Insoluble in water. Soluble in nitric acid but insoluble in ethanol.
Formula Weight 62.3
Percent Purity 99.9%
Grade Optical
Odor Odorless
Refractive Index 1.35-1.38
Density 3.148 g/mL
Assay (metals basis)
Chemical Name or Material Magnesium fluoride
Show More Show Less
compliance-icons
Product Identifier
  • Magnesium fluoride, Optical Grade
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Magnesium fluoride hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.