missing translation for 'onlineSavingsMsg'
Learn More

Magnesium fluoride, 99% (metals basis excluding Ca & Na), Ca+Na <1%

Product Code. 11368707
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368707 50 g 50g
11348707 250 g 250g
11358707 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368707 Supplier Thermo Scientific Alfa Aesar Supplier No. 035807.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7783-40-6 | F2Mg | 62.30 g/mol

Used in optics, space telescopes. Most commonly used on the surfaces of optical elements as inexpensive anti-reflective coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-40-6
Molecular Formula F2Mg
Molecular Weight (g/mol) 62.30
MDL Number MFCD00011108
InChI Key ORUIBWPALBXDOA-UHFFFAOYSA-L
IUPAC Name magnesium(2+) difluoride
SMILES [F-].[F-].[Mg++]

Specifications

Melting Point 1261°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99%, Ca+Na <1% (Metals basis excluding Ca and Na)
Solubility Information Sparingly soluble in water. Slightly soluble in dilute HNO3Insoluble in ethanol.
Formula Weight 62.3
Odor Odorless
Refractive Index 1.35-1.38
Density 3.148 g/cm3 at 20°C
Chemical Name or Material Magnesium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.