missing translation for 'onlineSavingsMsg'
Learn More

Magnesium fluoride, 99.9% (metals basis excluding Ca & Na), Ca+Na <1%

Product Code. 11397848
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11397848

Brand: Thermo Scientific Alfa Aesar 036391.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vacuum deposition | CAS: 7783-40-6 | F2Mg | 62.30 g/mol

Magnesium fluoride is used as Vacuum deposition, anti-reflective coatings, in optics and also used in space telescopes, glass industry and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-40-6
Molecular Formula F2Mg
Molecular Weight (g/mol) 62.30
MDL Number MFCD00011108
InChI Key ORUIBWPALBXDOA-UHFFFAOYSA-L
IUPAC Name magnesium(2+) difluoride
SMILES [F-].[F-].[Mg++]

Specifications

Melting Point 1261°C
Color White
Physical Form Vacuum Melted Lump
Quantity 25 g
Linear Formula MgF2
Merck Index 14,5665
Solubility Information Soluble in water(0.076g/L),nitric acid. Insoluble in ethanol.
Formula Weight 62.3
Percent Purity 99.9%
Odor Odorless
Refractive Index 1.35-1.38
Density 3.148 g/mL
Assay (metals basis excluding Ca and Na), Ca+Na <1%
Chemical Name or Material Magnesium fluoride
Mostrar más Mostrar menos

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado