missing translation for 'onlineSavingsMsg'
Learn More

Magnesium fluoride, 99.9% (metals basis excluding Ca & Na), Ca+Na <1%

Product Code. 11387848
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387848 100 g 100g
11397848 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387848 Supplier Thermo Scientific Alfa Aesar Supplier No. 036391.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Vacuum deposition | CAS: 7783-40-6 | F2Mg | 62.30 g/mol

Magnesium fluoride is used as Vacuum deposition, anti-reflective coatings, in optics and also used in space telescopes, glass industry and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-40-6
Molecular Formula F2Mg
Molecular Weight (g/mol) 62.30
MDL Number MFCD00011108
InChI Key ORUIBWPALBXDOA-UHFFFAOYSA-L
IUPAC Name magnesium(2+) difluoride
SMILES [F-].[F-].[Mg++]

Specifications

Melting Point 1261°C
Color White
Physical Form Vacuum Melted Lump
Quantity 100 g
Linear Formula MgF2
Merck Index 14,5665
Solubility Information Soluble in water(0.076g/L),nitric acid. Insoluble in ethanol.
Formula Weight 62.3
Percent Purity 99.9%
Odor Odorless
Refractive Index 1.35-1.38
Density 3.148 g/mL
Assay (metals basis excluding Ca and Na), Ca+Na <1%
Chemical Name or Material Magnesium fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.