missing translation for 'onlineSavingsMsg'
Learn More

Magnesium ethoxide, Mg 21-22%

Product Code. 11387847
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387847 50 g 50g
11397847 250 g 250g
11307857 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387847 Supplier Thermo Scientific Alfa Aesar Supplier No. 014264.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2414-98-4 | C4H10MgO2 | 114.427 g/mol

Magnesium ethoxide is used as olefin polymerization catalyst carrier for polypropylene, polyethylene and precision ceramic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2414-98-4
Molecular Formula C4H10MgO2
Molecular Weight (g/mol) 114.427
MDL Number MFCD00009060
InChI Key XDKQUSKHRIUJEO-UHFFFAOYSA-N
Synonym magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide
PubChem CID 164963
IUPAC Name magnesium;ethanolate
SMILES CC[O-].CC[O-].[Mg+2]

Specifications

Melting Point 270°C (decomposition)
Color White
Density 1.01g/mL
Odor Odorless
Linear Formula Mg(OCH2CH3)2
Quantity 50 g
UN Number UN3205
Beilstein 3678841
Sensitivity Air & Moisture Sensitive
Formula Weight 114.44
Concentration or Composition (by Analyte or Components) Mg 21 to 22%
Physical Form Granules
Chemical Name or Material Magnesium ethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.