missing translation for 'onlineSavingsMsg'
Learn More

Magnesium ethoxide, 97+%

Product Code. 10204784 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10204784 5 g Glass bottle 5g
10041564 100 g Glass bottle 100g
10674565 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10204784 Supplier Thermo Scientific Acros Supplier No. 397470050
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2414-98-4 | C4H10MgO2 | 114.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2414-98-4
Molecular Formula C4H10MgO2
Molecular Weight (g/mol) 114.43
MDL Number MFCD00009060
InChI Key XDKQUSKHRIUJEO-UHFFFAOYSA-N
Synonym magnesium ethoxide, magnesium ethanolate, magnesium ethylate, magnesium diethanolate, unii-o81sa5a7l6, magnesium diethoxide, magnesium 2+ bis ethoxide, ethanol, magnesium salt, magnesiumethoxide, magnesium ii ethoxide
PubChem CID 164963
IUPAC Name magnesium;ethanolate
SMILES CC[O-].CC[O-].[Mg+2]

Specifications

Melting Point 270.0°C
Color White to Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3CH2O)2Mg
Quantity 5 g
Solubility Information Solubility in water: reacts
Formula Weight 114.43
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Magnesium ethoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Magnesium ethoxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 1
  • Self-heating substance Category 1
Hazard Statement
  • H319-Causes serious eye irritation.
  • H251-Self-heating: may catch fire.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235+P410-Keep cool. Protect from sunlight.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.