missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Di Citrate, MP Biomedicals

Product Code. 11432066 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432066 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11432066 Supplier MP Biomedicals Supplier No. 0521225680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 3344-18-1
Molecular Formula C12H12Mg3O15
Quantity 100 g
Synonym trimagnesium 2+ dicitrate hydrate
InChI Key HZTVQLXIRHEMMB-UHFFFAOYSA-H
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Mg+2].[Mg+2].[Mg+2]
IUPAC Name trimagnesium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
Molecular Weight (g/mol) 469.128
PubChem CID 122198783
Formula Weight 703.36
Chemical Name or Material Magnesium Di Citrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.