missing translation for 'onlineSavingsMsg'
Learn More

Magnesium citrate hydrate, Mg ca 16% (dry wt.)

Product Code. 11384897
Change view
Click to view available options
Quantity:
1 kg
5 kg
17 kg
Unit Size:
17kg
1kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384897 1 kg 1kg
11394897 5 kg 5kg
11315949 17 kg 17kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384897 Supplier Thermo Scientific Alfa Aesar Supplier No. 041921.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3344-18-1 | C12H12Mg3O15 | 469.128 g/mol

Used as a laxative or dietary supplement

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3344-18-1
Molecular Formula C12H12Mg3O15
Molecular Weight (g/mol) 469.128
MDL Number MFCD09953657
InChI Key HZTVQLXIRHEMMB-UHFFFAOYSA-H
Synonym trimagnesium 2+ dicitrate hydrate
PubChem CID 122198783
IUPAC Name trimagnesium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[Mg+2].[Mg+2].[Mg+2]

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Merck Index 14,5663
Solubility Information Slightly soluble in water.
Formula Weight 451.13 (Anhydrous)
Concentration or Composition (by Analyte or Components) Mg ≈ 16% (dry wt.)
Chemical Name or Material Magnesium citrate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.