missing translation for 'onlineSavingsMsg'
Learn More

Magnesium citrate, dibasic hydrate

Product Code. 11384937
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384937 250 g 250g
11394937 1 kg 1kg
11304947 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384937 Supplier Thermo Scientific Alfa Aesar Supplier No. 041954.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119851-23-9 | C6H6MgO7 | 214.41 g/mol

Magnesium citrate is used to clean stool from the intestines before surgery. It is also used as a laxative for relief of constipation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119851-23-9
Molecular Formula C6H6MgO7
Molecular Weight (g/mol) 214.41
MDL Number MFCD00151706
InChI Key DIXGJWCZQHXZNR-UHFFFAOYNA-L
Synonym magnesium 2-carboxymethyl-2-hydroxysuccinate hydrate
PubChem CID 88316079
SMILES [Mg++].OC(=O)CC(O)(CC([O-])=O)C([O-])=O

Specifications

Physical Form Powder
Quantity 250 g
Merck Index 14,5663
Solubility Information Slightly soluble in water.
IUPAC Name magnesium(2+) 2-(carboxymethyl)-2-hydroxybutanedioate
Formula Weight 214.14 (Anhydrous)
Chemical Name or Material Magnesium citrate, dibasic hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.