missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chromate hydrate, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11348488
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348488 100 g 100g
11338488 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348488 Supplier Thermo Scientific Alfa Aesar Supplier No. 040503.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Corrosion inhibitor, pigment

Used as Corrosion inhibitor, pigment. As an ingredient in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16569-85-0
Molecular Formula CrMgO4
Molecular Weight (g/mol) 140.30
MDL Number MFCD00801009
InChI Key CRGGPIWCSGOBDN-UHFFFAOYSA-N
Synonym Magnesium chromate
IUPAC Name magnesium(2+) dioxochromiumbis(olate)
SMILES [Mg++].[O-][Cr]([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 100 g
UN Number UN2923
Solubility Information Soluble in water
Formula Weight 140.30 (Anhydrous)
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Magnesium chromate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.