missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride, ultra dry, 99.99% (metals basis)

Product Code. 11340078
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340078 5 g 5g
11330078 25 g 25g
11320078 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11340078 Supplier Thermo Scientific Alfa Aesar Supplier No. 042850.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol

Magnesium chloride is generally used as a Lewis acid catalyst. It is used as a flocculating agent, analytical reagent, anesthetic for cephalopods, and also in various magnesium cements, and ceramics. It is used for dust control, and road stabilization. It is used in fire extinguishers, polymerase chain reaction (PCR) and hydrogen storage. It is a precursor for magnesium metal and other magnesium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7786-30-3
Molecular Formula Cl2Mg
Molecular Weight (g/mol) 95.21
MDL Number MFCD00011106
InChI Key TWRXJAOTZQYOKJ-UHFFFAOYSA-L
Synonym magnesium chloride, magnesium chloride anhydrous, mgcl2, magnesium chloride, anhydrous, magnesium 2+ ion dichloride, chloromagnesite, magnesiumchlorid, magnesium ii chloride
PubChem CID 5360315
ChEBI CHEBI:6636
SMILES [Mg++].[Cl-].[Cl-]

Specifica

Melting Point 714°C
Color White to Yellow
Physical Form Powder
Quantity 5 g
Odor Odorless
Sensitivity Hygroscopic
Appearance White to yellow powder
Assay Percent Range 99.99% (Metal basis)
Density 2.32 g/cm3 at 20°C
Solubility Information Highly soluble in water with evolution of heat. Slightly soluble in acetone and pyridine.
IUPAC Name magnesium(2+) dichloride
Formula Weight 95.22
Chemical Name or Material Magnesium chloride, ultra dry
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.