missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number p-3257030
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7791-18-6
Cl2H12MgO6
203.30
MFCD00149781
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
This item is not returnable. View return policy
7791-18-6
Cl2H12MgO6
203.30
MFCD00149781
DHRRIBDTHFBPNG-UHFFFAOYSA-L
magnesium(2+) hexahydrate dichloride
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Utilized in the polymerase chain reactions (PCR). Used for dust control, ice control, and road stabilization. Employed in a variety of applications such as fertilizer, mineral supplement for animals, waste water treatment, artificial seawater, textiles, paper, fireproofing agents, cements, and fire extinguishers. Used in several medicines and topical or skin related applications. Acts as a coagulant in the preparation of tofu from soy milk. Acts as an anaesthetic for cephalopods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 117°C (decomposition)
Color White
Physical Form Crystalline Aggregates
Quantity 25 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Highly soluble in water and alcohol. Slightly soluble in acetone and pyridine.
Formula Weight 203.30 (95.22 Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White Crystalline Aggregates
Density 1.57 g/cm3 at 20°C
Chemical Name or Material Magnesium chloride hexahydrate, hexahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'