missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, MP Biomedicals™

Product Code. 11440532 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440532 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11440532 Supplier MP Biomedicals Supplier No. 02191421.5
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to prepare renaturation buffer. Magnesium Chloride Hexahydrate, MP Biomedicals is an inorganic salt that can be generated from the acid leach solution of magnesite tailings. It is a potential phase change material (PCM) for storing thermal energy in

Magnesium chloride hexahydrate (MgCl2.6H2O) has been used to prepare renaturation buffer that is employed for the renaturation of denatured peptidoglycan (PGN) hydrolytic enzymes. It may be used as a catalyst in the preparation of 3,4-dihydropyrimidin-2-(1H)-ones. The result of altering poly(vinyl alcohol) (PVA) films with magnesium chloride hexahydrate has been investigated. Its ability to reduce the flammability of cotton fabrics has also been studied.

Specifications

CAS 7791-18-6
Physical Form Crystals
Quantity 500 g
Molecular Formula Cl2H12MgO6
MDL Number MFCD00149781
Synonym Magnesium dichloride hexahydrate, Magnesium chloride hydrate, Magnesium chloride
Solubility Information Soluble in water (500mg/mL), yielding a clear, colorless solution.
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
IUPAC Name magnesium(2+) hexahydrate dichloride
Molecular Weight (g/mol) 203.30
Formula Weight 203.3
Percent Purity 99 to 102%
Grade ACS Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.