missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, (Crystalline),MP Biomedicals

Product Code. 11411911 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411911 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411911 Supplier MP Biomedicals Supplier No. 0219469880
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7791-18-6
Physical Form Crystalline
Quantity 100 g
Molecular Formula Cl2H12MgO6
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
IUPAC Name magnesium(2+) hexahydrate dichloride
Molecular Weight (g/mol) 203.30
Formula Weight 203.3
Chemical Name or Material Magnesium Chloride Hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.