missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate (Crystalline/Cell Culture)

Product Code. 11421911 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421911 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11421911 Supplier MP Biomedicals Supplier No. 0219469890
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity magnesium chloride hexahydrate designed for use as a cell culture reagent.

Specifications

CAS 7791-18-6
Physical Form Crystalline Powder
Quantity 500 g
Molecular Formula Cl2H12MgO6
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
IUPAC Name magnesium(2+) hexahydrate dichloride
Molecular Weight (g/mol) 203.30
Percent Purity 99.1%
Grade Cell Culture
Chemical Name or Material Magnesium Chloride Hexahydrate (Crystalline/Cell Culture)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.