missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10795921 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10795921 1 kg 1kg
10785921 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10795921 Supplier JT Baker Supplier No. 0162.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Meets ACS Specifications

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Boiling Point 1412°C (1013 hPa)
Melting Point 117°C
Quantity 1 kg
Assay Percent Range 99.0 to 102.0%
Density 1.57 g/cm³ (25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.