missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride Hexahydrate, 99.0-101.0%, (Crystalline/ACS Reagent Grade), MP Biomedicals

Product Code. 11460532 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460532 5 kg 5kg
11450532 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460532 Supplier MP Biomedicals Supplier No. 0219142105
  • ONLINE EXCLUSIVE PRICE
    £735.00
    £688.80 / 5kg
    Save £46.20
    6% Off
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7791-18-6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Physical Form Crystalline
Quantity 5 kg
Assay Percent Range 99.0-101.0%
Molecular Formula Cl2H12MgO6
Formula Weight 203.3
Percent Purity 99 to 102%
Grade ACS Reagent
Chemical Name or Material Magnesium Chloride Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.