missing translation for 'onlineSavingsMsg'
Learn More

Magnesium chloride hexahydrate, 97+%

Product Code. 11361639
Change view
Click to view available options
Quantity:
2 kg
Unit Size:
2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361639 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11361639 Supplier Thermo Scientific Alfa Aesar Supplier No. 012288.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7791-18-6 | Cl2H12MgO6 | 203.30 g/mol

It is utilized in the polymerase chain reactions (PCR). It is used for dust control, ice control, and road stabilization. It is employed in a variety of applications such as fertilizers, mineral supplements for animals, waste water treatment, artificial seawater, textiles, paper, fireproofing agents, cements, and fire extinguishers. Pharmaceutical preparations are used in several medicines and topical or skin related applications. It acts as a coagulant in the preparation of tofu from soy milk.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Melting Point 117°C (decomposition)
Color White
Physical Form Flakes
Quantity 2 kg
Assay Percent Range >98%
Solubility Information Highly soluble in water and alcohol. Slightly soluble in acetone and pyridine.
Formula Weight 203.30 (95.22anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White flakes
Density 1.57
Chemical Name or Material Magnesium chloride hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.